(R)-3-Amino-3-(4-cyanophenyl)propionic acid 95%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Amino-3-(4-cyanophenyl)propanoic acid

A311355-100MG A311355-250MG A311355-1G
103814-660EA 105 USD
103814-660 77141-690 77141-692 77306-048
(R)-3-Amino-3-(4-cyanophenyl)propionic acid 95%
(R)-3-Amino-3-(4-cyanophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
Boiling Pt: 395.4 °C (760 mm Hg)
Melting Pt: 240…242 °C
Flash Pt: 192.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04113665
CAS Number: 738606-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR