(R)-3-(Methoxymethyl)pyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A145756-100MG A145756-250MG
103803-582EA 134 USD
103803-582 77150-248 77150-250
(R)-3-(Methoxymethyl)pyrrolidine hydrochloride 95%
(R)-3-(Methoxymethyl)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29037448
CAS Number: 955400-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR