Boc-L-β-homovaline 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-β-leucine, Boc-β-HoVal-OH, Boc-β-HomoVal-OH, (R)-3-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid

A102083-100MG A102083-250MG A102083-5G
103796-286EA 30 USD
103796-286 103821-980 76994-560 76994-562 77308-866
Boc-L-β-homovaline 97%
Boc-L-beta-homovaline

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
Melting Pt: 115…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01076232
CAS Number: 183990-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR