(R)-3-((tert-Butoxycarbonyl)amino)-4-(1H-indol-3-yl)butanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-β-HoTrp-OH

A787059-1G A787059-250MG
103841-000EA 733 USD
103841-000 76845-252 77159-100
(R)-3-((tert-Butoxycarbonyl)amino)-4-(1H-indol-3-yl)butanoic acid 95%
(R)-3-((tert-Butoxycarbonyl)amino)-4-(1H-indol-3-yl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₄
MW: 318.36 g/mol
Boiling Pt: 550.1±45.0 °C (760 mm Hg)
Flash Pt: 286.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08276227
CAS Number: 1073269-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR