(R)-3-(Fmoc-amino)-3-phenylpropionic acid 95%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylpropanoic acid

A435193-1G A435193-5G A435193-250MG
103827-590EA 46 USD
103827-590 103838-658 76842-376 76842-378 77004-226
(R)-3-(Fmoc-amino)-3-phenylpropionic acid 95%
(R)-3-(Fmoc-amino)-3-phenylpropionic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₄
Boiling Pt: 617.0±50.0 °C (760 mm Hg)
Melting Pt: 181.7 °C
Flash Pt: 326.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01311783
CAS Number: 220498-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR