(R)-2-Phenylpyrrolidine 97%

Supplier: Ambeed
Warning

A215138-100MG A215138-250MG A215138-1G
103818-460EA 253 USD
103818-460 77139-722 77139-724 77303-398
(R)-2-Phenylpyrrolidine 97%
(R)-2-Phenylpyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.21 g/mol
Boiling Pt: 237 °C (760 mm Hg)
Flash Pt: 98.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06762513
CAS Number: 56523-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR