(R)-(-)-2-Octanol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-2-Hydroxy octane, (R)-2-Octanol, L(-)-2-Octanol, (R)-(-)-Hexyl methyl carbinol, (R)-(-)-Methyl hexyl carbinol

A466919-100G A466919-10G A466919-1G A466919-5G
103827-988EA 5 USD
103827-988 103839-062 76774-260 76774-262 76842-972 76842-974
(R)-(-)-2-Octanol 98%
(R)-(-)-2-Octanol

Precaución: for Research Use Only

Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 175 °C (760 mm Hg)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00064284
CAS Number: 5978-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR