(R)-(-)-2-Methylpyrrolidine 97%

Supplier: Ambeed
Danger

Synonyms: (R)-2-Methylpyrrolidine

A201528-100MG A201528-10G A201528-250MG A201528-1G A201528-25G A201528-5G
103817-306EA 35 USD
103817-306 103847-946 76873-422 76873-424 76873-426 77086-020 77086-022 77086-024
(R)-(-)-2-Methylpyrrolidine 97%
(R)-(-)-2-Methylpyrrolidine

Precaución: for Research Use Only

Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 100.2 °C (760 mm Hg)
Flash Pt: 7.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07783026
CAS Number: 41720-98-3
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR