(R)-(-)-2-Methylpiperazine 97%

Supplier: Ambeed
Danger

A494420-25G A494420-5G
103839-320EA 6 USD
103839-320 76843-286 77307-590
(R)-(-)-2-Methylpiperazine 97%
(R)-(-)-2-Methylpiperazine

Precaución: for Research Use Only

Formula: C₅H₁₂N₂
Boiling Pt: 155 °C (760 mm Hg)
Melting Pt: 91…93 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00192317
CAS Number: 75336-86-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR