(R)-(+)-2-Methyl-2-propanesulfinamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2-methylpropane-2-sulphinamide, R)-(+)-tert-Butyl sulphinamide, (R)-2-Methyl-2-propanesulphinamide, (R)-(+)-tert-Butylsulphinamide, (R)-(+)-t-Butylsulphinamide
A348213-25G
A348213-100G
A348213-10G
A348213-1G
A348213-500G
A348213-5G
A348213-1000G
103836-976EA
8
USD
103836-976
76840-054
77003-222
77003-224
77003-226
77003-228
77306-594
77601-554
(R)-(+)-2-Methyl-2-propanesulfinamide 98%
(R)-(+)-2-Methyl-2-propanesulfinamide
Precaución: for Research Use Only
Formula:
C₄H₁₁NOS MW: 121.20 g/mol Melting Pt: 103…107 °C Flash Pt: 86.827 °C Storage Temperature: Ambient |
MDL Number:
MFCD05861479 CAS Number: 196929-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...