(R)-2-Hydroxy-4-methylpentanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: D-α-Hydroxyisocaproic acid, (R)-Leucic acid

A318657-1G A318657-10G A318657-250MG A318657-25G A318657-5G
103825-182EA 34 USD
103825-182 103836-116 76838-970 77141-760 77141-762 77306-186 77306-188
(R)-2-Hydroxy-4-methylpentanoic acid 95%
(R)-2-Hydroxy-4-methylpentanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.159 g/mol
Boiling Pt: 251.3 °C (760 mm Hg)
Melting Pt: 76…80 °C
Flash Pt: 120.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00211263
CAS Number: 20312-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR