(R)-(-)-2-Hydroxyisovaleric acid 97%
Supplier: Ambeed
|
Warning
|
A291232-100MG
A291232-10G
A291232-250MG
A291232-1G
A291232-25G
A291232-5G
103813-900EA
8
USD
103813-900
103835-040
76693-230
76693-232
76693-234
76837-628
76837-630
76837-632
(R)-(-)-2-Hydroxyisovaleric acid 97%
(R)-(-)-2-Hydroxyisovaleric acid
Precaución: for Research Use Only
Formula:
C₅H₁₀O₃ Melting Pt: 64…67 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066442 CAS Number: 17407-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...