(R)-(-)-2-Chloro-1-phenylethanol 98%

Supplier: Ambeed
Warning

A235309-1G A235309-25G A235309-5G
103811-002EA 17 USD
103811-002 103850-766 77304-008 77304-010 77304-012
(R)-(-)-2-Chloro-1-phenylethanol 98%

Precaución: for Research Use Only

Formula: C₈H₉ClO
Boiling Pt: 254.9 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077691
CAS Number: 56751-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR