(R)-2-Benzyl 1-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate 95%

Supplier: Ambeed
Warning

A316741-100MG A316741-10G A316741-250MG A316741-1G A316741-25G A316741-5G
103825-114EA 68 USD
103825-114 103836-044 76816-980 76816-982 76816-984 76838-878 76838-880 76838-882
(R)-2-Benzyl 1-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate 95%
(R)-2-Benzyl 1-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁NO₅
MW: 319.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14635665
CAS Number: 400626-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR