(R)-2-Aminopent-4-ynoic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-D-Propargyl-Gly-OH·HCl

A440101-1G A440101-10G
103827-654EA 44 USD
103827-654 103838-720 76842-478 76877-370
(R)-2-Aminopent-4-ynoic acid hydrochloride 95%
(R)-2-Aminopent-4-ynoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈ClNO₂
MW: 149.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09263338
CAS Number: 87205-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR