D-Propargylglycine 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Pra-OH, (R)-2-Aminopent-4-ynoic acid, (R)-alpha-Propargylglycine

A135941-10G A135941-250MG A135941-5G A135941-1G
103802-248EA 107 USD
103802-248 77095-204 77095-206 77298-834 77315-132
D-Propargylglycine 98%
D-Propargylglycine

Precaución: for Research Use Only

Formula: C₅H₇NO₂
Boiling Pt: 272.1 °C (760 mm Hg)
Flash Pt: 118.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237407
CAS Number: 23235-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR