(R)-2-Aminobutanamide hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-2-Aminobutanamide HCl, (R)-2-Aminobutyramide hydrochloride, (2R)-2-Aminobutanamide Hydrochloride, (R)-(–)-2-Aminobutanamide hydrochloride

A121437-100G A121437-25G A121437-1G A121437-5G A121437-10G
103800-074EA 7 USD
103800-074 77136-498 77136-500 77312-196 77312-198 77614-946
(R)-2-Aminobutanamide hydrochloride 95%
(R)-2-Aminobutanamide hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClN₂O
MW: 138.59 g/mol
Melting Pt: 258…262 °C
Flash Pt: 119.6 °C
Storage Temperature: Ambient
MDL Number: MFCD09265126
CAS Number: 103765-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR