(R)-2-Amino-5-methylhexanoic acid 97%
Supplier: Ambeed
|
Warning
|
A212628-1G
A212628-5G
A212628-100MG
A212628-250MG
A212628-25G
103818-220EA
54
USD
103818-220
103848-916
77087-384
77087-386
77139-652
77139-654
77139-656
(R)-2-Amino-5-methylhexanoic acid 97%
(R)-2-Amino-5-methylhexanoic acid
Precaución: for Research Use Only
Formula:
C₇H₁₅NO₂ MW: 145.2 g/mol Boiling Pt: 243.4 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD06796740 CAS Number: 138751-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...