(R)-2-amino-3-cyclohexylpropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-3-cyclohexylpropanoic acid

A555997-1G A555997-25G A555997-5G A555997-100G A555997-10G A555997-250MG
103828-716EA 6 USD
103828-716 103839-852 76843-944 76843-946 76843-948 77116-964 77116-966 77116-968
(R)-2-amino-3-cyclohexylpropanoic acid 95%
(R)-2-amino-3-cyclohexylpropanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Boiling Pt: 307.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00153485
CAS Number: 58717-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR