(R)-2-Amino-2-phenylacetamide 98%

Supplier: Ambeed
Warning

Synonyms: D(-)-Phenylglycinamide, H-D-Phg-NH₂·HCl, H-D-Phg-NH2

A390478-10G A390478-5G A390478-100G A390478-1G
103837-906EA 12 USD
103837-906 76817-772 76841-296 77572-184 77672-942
(R)-2-Amino-2-phenylacetamide 98%
(R)-2-Amino-2-phenylacetamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.181 g/mol
Boiling Pt: 322.8 °C (760 mm Hg)
Melting Pt: 125…129 °C
Flash Pt: 149 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06799064
CAS Number: 6485-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR