(R)-2-Amino-2-(tetrahydro-2H-pyran-4-yl)acetic acid 97%

Supplier: Ambeed
Warning

A190928-1G A190928-100MG A190928-250MG
103816-334EA 158 USD
103816-334 103846-940 77084-770 77111-756 77111-758
(R)-2-Amino-2-(tetrahydro-2H-pyran-4-yl)acetic acid 97%
(R)-2-Amino-2-(tetrahydro-2H-pyran-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₃
MW: 159.18 g/mol
Boiling Pt: 316.7±17.0 °C (760 mm Hg)
Melting Pt: 3>320…334 °C
Storage Temperature: Ambient
MDL Number: MFCD01631205
CAS Number: 475649-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR