D(-)-2-(2,5-Dihydrophenyl)glycine 97%

Supplier: Ambeed
Warning

A215318-100G A215318-500G A215318-25G A215318-5G
103849-182EA 13 USD
103849-182 76770-662 76770-664 77303-406 77303-408
D(-)-2-(2,5-Dihydrophenyl)glycine 97%
D(-)-2-(2,5-Dihydrophenyl)glycine

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
Boiling Pt: 279.4 °C (760 mm Hg)
Melting Pt: 280 °C (760 mm Hg)
Flash Pt: 122.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00137746
CAS Number: 26774-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR