4-Fluoro-D-2-phenylglycine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Phg(4-F)-OH, (R)-2-Amino-2-(4-fluorophenyl)acetic Acid
A111711-250MG
A111711-1G
A111711-5G
A111711-10G
A111711-25G
103797-888EA
36
USD
103797-888
103823-706
77245-726
77284-158
77284-160
77745-825
77745-826
4-Fluoro-D-2-phenylglycine 95%
4-Fluoro-D-2-phenylglycine
Precaución: for Research Use Only
Formula:
C₈H₈FNO₂ Boiling Pt: 289.9 °C (760 mm Hg) Melting Pt: 300 °C Flash Pt: 129.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042727 CAS Number: 93939-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...