(R)-2-Acetoxy-2-phenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-O-Acetylmandelic acid

A372846-10G A372846-25G A372846-100G A372846-500G
103837-538EA 7 USD
103837-538 76694-358 76840-800 77594-112 77601-650
(R)-2-Acetoxy-2-phenylacetic acid 98%
(R)-2-Acetoxy-2-phenylacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 317.8 °C (760 mm Hg)
Melting Pt: 97…99 °C
Flash Pt: 125 °C
Storage Temperature: Freezer
MDL Number: MFCD00004249
CAS Number: 51019-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR