(R)-2-(Benzylamino)propan-1-ol 98%
Supplier: Ambeed
|
Warning
|
A484417-100MG
A484417-250MG
A484417-25G
103828-164EA
39
USD
103828-164
103839-238
77241-356
77241-358
77746-997
(R)-2-(Benzylamino)propan-1-ol 98%
(R)-2-(Benzylamino)propan-1-ol
Precaución: for Research Use Only
Formula:
C₁₀H₁₅NO MW: 165.23 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20441630 CAS Number: 74609-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...