(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid 98% (>80% EE)
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen, (2R)-2-(6-Methoxynaphth-2-yl)propionic acid, R-Naproxen, 2-[(1R)-1-Carboxyethyl]-6-methoxynaphthalene
A106813-100MG
A106813-10G
A106813-250MG
A106813-25G
A106813-1G
A106813-5G
103797-080EA
44
USD
103797-080
103822-838
76780-864
76780-866
76780-868
76780-870
77309-690
77309-692
(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid 98% (>80% EE)
(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₄O₃ MW: 230.259 g/mol Boiling Pt: 403.9 °C (760 mm Hg) Melting Pt: 156…158 °C Flash Pt: 154.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00870716 CAS Number: 23979-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...