(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid 96%
Supplier: Ambeed
|
Warning
|
A101600-10G
A101600-250MG
A101600-25G
A101600-500G
A101600-1G
A101600-5G
A101600-100G
103796-042EA
18
USD
103796-042
103821-730
76780-512
76780-514
76780-516
76780-518
77282-768
77282-770
77666-472
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid 96%
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₇N₃O₆ Melting Pt: 205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792472 CAS Number: 62893-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...