(R)-(1-BOC-pyrrolidin-3-yl)-acetic acid 97%

Supplier: Ambeed
Danger

A602926-1G A602926-10G
103828-986EA 10 USD
103828-986 103840-126 76844-286 77104-068 77104-070
(R)-(1-BOC-pyrrolidin-3-yl)-acetic acid 97%
(R)-(1-BOC-pyrrolidin-3-yl)-acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
Boiling Pt: 357.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD05861547
CAS Number: 204688-60-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR