Boc-D-norvaline 95%

Supplier: Ambeed
Warning

A127935-1G A127935-5G A127935-100G
103801-040EA 9 USD
103801-040 77313-314 77313-316 77611-112
Boc-D-norvaline 95%
Boc-D-norvaline

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
Boiling Pt: 348.3 °C (760 mm Hg)
Flash Pt: 164.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00155638
CAS Number: 57521-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR