Boc-D-propargylglycine 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-2-propargyl-D-glycine, Boc-D-Pra-OH, (R)-2-((tert-Butoxycarbonyl)amino)pent-4-ynoic acid, (R)-N-BOC-Propargylglycine

A587769-10G A587769-250MG A587769-5G A587769-25G
103828-904EA 28 USD
103828-904 103840-050 76819-310 76819-312 76844-194 77307-894
Boc-D-propargylglycine 97%
Boc-D-propargylglycine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₄
Boiling Pt: 365.2 °C (760 mm Hg)
Melting Pt: 90…94 °C
Flash Pt: 157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320886
CAS Number: 63039-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR