Boc-D-Ser(Me)-OH 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-methoxypropanoic acid, Boc-O-Methyl-D-serine

A122750-100G A122750-10G A122750-250MG A122750-25G A122750-1G A122750-5G
103800-276EA 7 USD
103800-276 76797-466 76797-468 76797-470 76797-472 77312-390 77312-392
Boc-D-Ser(Me)-OH 97%
Boc-D-Ser(Me)-OH

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.2 g/mol
Boiling Pt: 355.8 °C (760 mm Hg)
Flash Pt: 169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08063987
CAS Number: 86123-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR