Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-Val(3-OH)-OH, (R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid, 3-Hydroxy-D-valine, N-BOC protected
A376609-100MG
A376609-250MG
A376609-1G
A376609-5G
103826-586EA
338
USD
103826-586
76724-838
76724-840
76840-916
77879-819
Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid 95%
Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₅ Boiling Pt: 391.1±37.0 °C (760 mm Hg) Melting Pt: 113…127 °C Flash Pt: 190.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02682576 CAS Number: 288159-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...