N-Boc-2,4,5-trifluoro-D-phenylalanine 96%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(2,4,5-trifluorophenyl)propanoic acid

A100497-100MG A100497-250MG A100497-1G
103795-478EA 21 USD
103795-478 103821-172 76686-996 76686-998 77296-720
N-Boc-2,4,5-trifluoro-D-phenylalanine 96%
N-Boc-2,4,5-trifluoro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₆F₃NO₄
MW: 319.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659118
CAS Number: 486460-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR