Boc-D-cyclopropylglycine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-(Boc-amino)-cyclopropylacetic acid, (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclopropylacetic acid, Boc-(R)-cyclopropylglycine

A188102-250MG A188102-25G A188102-1G A188102-5G
103816-074EA 73 USD
103816-074 103846-664 76690-760 76690-762 77084-390 77084-392
Boc-D-cyclopropylglycine 97%
Boc-D-cyclopropylglycine

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.246 g/mol
Boiling Pt: 364.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06659124
CAS Number: 609768-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR