Benzyl (R)-(-)-glycidyl ether 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2-((Benzyloxy)methyl)oxirane
A120722-10G
A120722-25G
A120722-1G
A120722-5G
A120722-100G
103799-986EA
5
USD
103799-986
76657-844
76657-846
77297-786
77312-094
77592-344
Benzyl (R)-(-)-glycidyl ether 98%
Benzyl (R)-(-)-glycidyl ether
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00068409 CAS Number: 14618-80-5 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...