(R)-2-(((Benzyloxy)carbonyl)amino)pent-4-enoic acid 97%

Supplier: Ambeed
Warning

A298801-5G A298801-100MG A298801-250MG
103814-196EA 17 USD
103814-196 76827-834 77885-795 77885-796
(R)-2-(((Benzyloxy)carbonyl)amino)pent-4-enoic acid 97%
(R)-2-(((Benzyloxy)carbonyl)amino)pent-4-enoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₄
Boiling Pt: 445.0±45.0 °C (760 mm Hg)
Flash Pt: 222.9 °C
Storage Temperature: Ambient
MDL Number: MFCD02094549
CAS Number: 127474-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR