Fmoc-Cys-OH 98%

Supplier: Ambeed
Warning

A566683-100MG A566683-250MG A566683-25G A566683-1G A566683-5G
103828-778EA 17 USD
103828-778 103839-918 76830-684 76830-686 76830-688 76844-030 76844-032
Fmoc-Cys-OH 98%
Fmoc-Cys-OH
Formula: C₁₈H₁₇NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD03788178
CAS Number: 135248-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR