(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopentylpropanoic acid 98%

Supplier: Ambeed
Warning

A101642-100MG A101642-250MG A101642-1G
103796-056EA 97 USD
103796-056 103821-752 77008-336 77008-338 77297-178
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopentylpropanoic acid 98%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopentylpropanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₅NO₄
MW: 379.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094883
CAS Number: 1262802-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR