Fmoc-cys(Bzl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(benzylthio)propanoic acid, N-Fmoc-S-benzyl-L-cysteine

A133187-1G A133187-5G A133187-10G
103801-840EA 6 USD
103801-840 77298-612 77314-464 77747-951
Fmoc-cys(Bzl)-OH 95%
Fmoc-cys(Bzl)-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄S
MW: 433.522 g/mol
Boiling Pt: 663.2 °C (760 mm Hg)
Flash Pt: 354.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065633
CAS Number: 53298-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR