Fmoc-(R)-2-(4-pentenyl)alanine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methylhept-6-enoic acid

A237821-250MG A237821-100MG
103811-178EA 169 USD
103811-178 103850-960 77593-528 77672-126
Fmoc-(R)-2-(4-pentenyl)alanine 97%
Fmoc-(R)-2-(4-pentenyl)alanine

Precaución: Inert atmosphere, store in freezer, under –20 °C.

Formula: C₂₃H₂₅NO₄
Boiling Pt: 591.7 °C (760 mm Hg)
Flash Pt: 311.671 °C
Storage Temperature: Freezer
MDL Number: MFCD09879889
CAS Number: 288617-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR