(R)-2-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)methyl)-3-methylbutanoic acid 98%

Supplier: Ambeed
Warning

A136562-250MG A136562-1G A136562-100MG
103792-152EA 75 USD
103792-152 103799-358 103802-320 76984-018 77298-860 77885-161
(R)-2-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)methyl)-3-methylbutanoic acid 98%
(R)-2-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)methyl)-3-methylbutanoic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD07372882
CAS Number: 501331-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR