(R)-1-tert-Butyl-3-methyl piperazine-1,3-dicarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate
A185926-1G
A185926-25G
A185926-5G
A185926-100G
A185926-10G
A185926-250MG
103815-894EA
7
USD
103815-894
103846-468
77084-114
77084-116
77084-118
77097-014
77097-016
77097-018
(R)-1-tert-Butyl-3-methyl piperazine-1,3-dicarboxylate 95%
(R)-1-tert-Butyl-3-methyl piperazine-1,3-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₂₀N₂O₄ MW: 244.2875 g/mol Boiling Pt: 321.3 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD04115325 CAS Number: 438631-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...