Ethyl (R)-1-(tert-butoxycarbonyl)-3-piperidinecarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl (R)-1-Boc-3-piperidinecarboxylate, (R)-1-(tert-Butoxycarbonyl)nipecotic Acid Ethyl Ester, (R)-1-Boc-3-piperidinecarboxylic Acid Ethyl Ester, (R)-1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid Ethyl Ester, (R)-1-Boc-nipecotic Acid Ethyl Ester, (R)-1-tert-Butyl 3-ethyl piperidine-1,3-dicarboxylate
A442658-1G
A442658-5G
A442658-250MG
103827-688EA
30
USD
103827-688
103838-758
76842-528
76842-530
77034-032
Ethyl (R)-1-(tert-butoxycarbonyl)-3-piperidinecarboxylate 95%
Ethyl (R)-1-(tert-butoxycarbonyl)-3-piperidinecarboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₂₃NO₄ MW: 257.33 g/mol Boiling Pt: 323.9 °C (760 mm Hg) Melting Pt: 36…40 °C Flash Pt: 149.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD07374389 CAS Number: 194726-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...