(R)-1-Phenylethyl (5-(4-bromophenyl)-3-methylisoxazol-4-yl)carbamate 97%

Supplier: Ambeed
Warning

A154940-250MG A154940-5G
103804-694EA 84 USD
103804-694 103842-890 77247-422 77247-424 77318-624
(R)-1-Phenylethyl (5-(4-bromophenyl)-3-methylisoxazol-4-yl)carbamate 97%
(R)-1-Phenylethyl (5-(4-bromophenyl)-3-methylisoxazol-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₁₇BrN₂O₃
MW: 401.25 g/mol
Boiling Pt: 484.2±45.0 °C (760 mm Hg)
Flash Pt: 246.6 °C
Storage Temperature: Freezer
MDL Number: MFCD25977339
CAS Number: 1228690-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR