(R)-β-Phenylalaninol 95%

Supplier: Ambeed
Danger

Synonyms: (R)-3-Amino-3-phenylpropan-1-ol, (R)-1-Phenyl-3-propanolamine, (R)-3-Amino-3-phenyl-1-propanol

A158407-100MG A158407-1G A158407-25G A158407-5G A158407-250MG
103805-128EA 17 USD
103805-128 103843-338 77247-568 77300-328 77300-330 77300-332 77615-284
(R)-β-Phenylalaninol 95%
(R)-β-Phenylalaninol

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.2056 g/mol
Flash Pt: 131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01311791
CAS Number: 170564-98-4
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR