(R)-1-Methylpiperidin-3-amine 97%

Supplier: Ambeed
Danger

A337754-1G A337754-100MG A337754-10G A337754-250MG A337754-5G
103825-754EA 51 USD
103825-754 103836-726 76839-742 76989-372 76989-374 76989-376 77306-508
(R)-1-Methylpiperidin-3-amine 97%
(R)-1-Methylpiperidin-3-amine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
Boiling Pt: 136.5±8.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD17015901
CAS Number: 1001353-92-9
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR