(R)-(+)-1-Boc-3-methylpiperazine 97%

Supplier: Ambeed
Warning

A110889-100G A110889-10G A110889-25G A110889-1G A110889-5G A110889-500G
103797-746EA 5 USD
103797-746 103823-554 76765-802 76765-804 76765-806 77284-068 77310-350 77623-918
(R)-(+)-1-Boc-3-methylpiperazine 97%
(R)-(+)-1-Boc-3-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 268.7 °C (760 mm Hg)
Melting Pt: 40…45 °C
Flash Pt: 106.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683205
CAS Number: 163765-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR