(R)-(-)-3-Amino-1-boc-piperidine 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl (3R)-3-aminopiperidine-1-carboxylate, (R)-1-Boc-3-Aminopiperidine

A104935-100G A104935-10G A104935-25G A104935-1G A104935-5G
103796-800EA 9 USD
103796-800 103822-526 76765-440 76765-442 76765-444 77283-306 77309-390
(R)-(-)-3-Amino-1-boc-piperidine 97%
(R)-(-)-3-Amino-1-Boc-piperidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Flash Pt: 121.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094717
CAS Number: 188111-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR