tert-Butyl (3R)-3-(methylamino)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (R)-3-(methylamino)pyrrolidine-1-carboxylate

A898773-1G A898773-25G A898773-5G A898773-250MG
103841-342EA 87 USD
103841-342 76845-602 76845-604 76845-606 77019-900
tert-Butyl (3R)-3-(methylamino)pyrrolidine-1-carboxylate 97%
tert-Butyl (3R)-3-(methylamino)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09752961
CAS Number: 199336-83-9
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR