(R)-1-Boc-2-Ethylpiperazine 97%

Supplier: Ambeed
Warning

A103777-100MG A103777-250MG
103796-612EA 36 USD
103796-612 103822-322 77093-618 77093-620
(R)-1-Boc-2-Ethylpiperazine 97%
(R)-1-Boc-2-ethylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3100 g/mol
Boiling Pt: 286.7 °C (760 mm Hg)
Flash Pt: 127.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411920
CAS Number: 393781-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR